Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGTACCAGTGGCCATGCTGGTGC[C/T]TAGGTCAGGGGAGGCGGTGGCGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601333 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PROCA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PROCA1 - protein interacting with cyclin A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304949.1 | 731 | Missense Mutation | AGC,GGC | S,G 164 | NP_001291878.1 | |
NM_001304951.1 | 731 | Missense Mutation | AGC,GGC | S,G 57 | NP_001291880.1 | |
NM_001304952.1 | 731 | Missense Mutation | AGC,GGC | S,G 175 | NP_001291881.1 | |
NM_001304953.1 | 731 | Missense Mutation | AGC,GGC | S,G 101 | NP_001291882.1 | |
NM_001304954.1 | 731 | Missense Mutation | AGC,GGC | S,G 101 | NP_001291883.1 | |
NM_152465.2 | 731 | Missense Mutation | AGC,GGC | S,G 173 | NP_689678.1 | |
XM_005257927.4 | 731 | Missense Mutation | AGC,GGC | S,G 201 | XP_005257984.1 | |
XM_006721723.3 | 731 | Missense Mutation | AGC,GGC | S,G 198 | XP_006721786.1 | |
XM_011524398.2 | 731 | Missense Mutation | AGC,GGC | S,G 192 | XP_011522700.1 | |
XM_011524399.2 | 731 | Missense Mutation | AGC,GGC | S,G 164 | XP_011522701.1 | |
XM_011524400.2 | 731 | Missense Mutation | AGC,GGC | S,G 147 | XP_011522702.1 | |
XM_011524401.1 | 731 | Missense Mutation | AGC,GGC | S,G 101 | XP_011522703.1 | |
XM_017024237.1 | 731 | Missense Mutation | AGC,GGC | S,G 101 | XP_016879726.1 |
SUPT6H - SPT6 homolog, histone chaperone | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |