Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGGGATACAATGGCCGGCTCAGT[A/C]GCCTTTAAAGCTGGTGGGAAGTAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611358 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPRXP4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DPRXP4 - divergent-paired related homeobox pseudogene 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF135 - ring finger protein 135 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184992.1 | Intron | NP_001171921.1 | ||||
NM_032322.3 | Intron | NP_115698.3 | ||||
NM_197939.1 | Intron | NP_922921.1 | ||||
XM_005258043.4 | Intron | XP_005258100.1 | ||||
XM_017025223.1 | Intron | XP_016880712.1 |