Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCATAAGCCCCGGGGGGCTCTGAT[A/G]TCATTGAATTAGGGGCCTCCTTTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601132 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KSR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KSR1 - kinase suppressor of ras 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014238.1 | Intron | NP_055053.1 | ||||
XM_006722154.3 | Intron | XP_006722217.1 | ||||
XM_011525429.2 | Intron | XP_011523731.1 | ||||
XM_011525430.2 | Intron | XP_011523732.1 | ||||
XM_011525431.2 | Intron | XP_011523733.1 | ||||
XM_017025267.1 | Intron | XP_016880756.1 | ||||
XM_017025268.1 | Intron | XP_016880757.1 | ||||
XM_017025269.1 | Intron | XP_016880758.1 | ||||
XM_017025270.1 | Intron | XP_016880759.1 | ||||
XM_017025271.1 | Intron | XP_016880760.1 | ||||
XM_017025272.1 | Intron | XP_016880761.1 | ||||
XM_017025273.1 | Intron | XP_016880762.1 | ||||
XM_017025274.1 | Intron | XP_016880763.1 | ||||
XM_017025275.1 | Intron | XP_016880764.1 | ||||
XM_017025276.1 | Intron | XP_016880765.1 | ||||
XM_017025277.1 | Intron | XP_016880766.1 | ||||
XM_017025278.1 | Intron | XP_016880767.1 | ||||
XM_017025279.1 | Intron | XP_016880768.1 | ||||
XM_017025280.1 | Intron | XP_016880769.1 |