Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCATCTGGGCCTTTCTGTCTTGTT[C/T]AACCTATGCATTCCAGTACAGAAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616178 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C17orf102 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C17orf102 - chromosome 17 open reading frame 102 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207454.2 | 1802 | UTR 3 | NP_997337.2 |
TMEM132E - transmembrane protein 132E | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |