Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGATGTTGGGCTTGTGACTGTAA[A/T]TATCACTTATCTGGTTGATGTCTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176887 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTPN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PTPN2 - protein tyrosine phosphatase, non-receptor type 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001207013.1 | 1644 | Intron | NP_001193942.1 | |||
NM_001308287.1 | 1644 | UTR 3 | NP_001295216.1 | |||
NM_002828.3 | 1644 | UTR 3 | NP_002819.2 | |||
NM_080422.2 | 1644 | Intron | NP_536347.1 | |||
NM_080423.2 | 1644 | Intron | NP_536348.1 | |||
XM_005258124.3 | 1644 | UTR 3 | XP_005258181.1 | |||
XM_005258125.3 | 1644 | Intron | XP_005258182.1 | |||
XM_011525705.2 | 1644 | UTR 3 | XP_011524007.1 | |||
XM_011525706.2 | 1644 | UTR 3 | XP_011524008.1 | |||
XM_011525707.2 | 1644 | UTR 3 | XP_011524009.1 | |||
XM_017025884.1 | 1644 | Intron | XP_016881373.1 | |||
XM_017025885.1 | 1644 | UTR 3 | XP_016881374.1 | |||
XM_017025886.1 | 1644 | UTR 3 | XP_016881375.1 | |||
XM_017025887.1 | 1644 | Intron | XP_016881376.1 | |||
XM_017025888.1 | 1644 | Intron | XP_016881377.1 |