Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTCTGGGATAAAAGTGAAAGTGGT[A/G]ATTGGCATTGTACTGCTAGCTGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609263 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEH1L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SEH1L - SEH1 like nucleoporin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013437.1 | 444 | Missense Mutation | AAT,GAT | N,D 46 | NP_001013455.1 | |
NM_031216.3 | 444 | Missense Mutation | AAT,GAT | N,D 46 | NP_112493.2 | |
XM_005258152.4 | 444 | Missense Mutation | AAT,GAT | N,D 46 | XP_005258209.1 | |
XM_011525742.2 | 444 | Missense Mutation | AAT,GAT | N,D 46 | XP_011524044.1 | |
XM_011525743.2 | 444 | Missense Mutation | AAT,GAT | N,D 46 | XP_011524045.1 | |
XM_011525744.1 | 444 | Missense Mutation | AAT,GAT | N,D 46 | XP_011524046.1 | |
XM_011525745.2 | 444 | UTR 5 | XP_011524047.1 | |||
XM_011525746.2 | 444 | UTR 5 | XP_011524048.1 | |||
XM_017026025.1 | 444 | Missense Mutation | AAT,GAT | N,D 46 | XP_016881514.1 | |
XM_017026026.1 | 444 | UTR 5 | XP_016881515.1 | |||
XM_017026027.1 | 444 | UTR 5 | XP_016881516.1 |