Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGAATGAGGTGGTGTTTGCCTCC[C/T]TCAGGTCTTTGACTTGGGGCTATAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607397 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MC2R PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MC2R - melanocortin 2 receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000529.2 | 2244 | UTR 3 | NP_000520.1 | |||
NM_001291911.1 | 2244 | UTR 3 | NP_001278840.1 | |||
XM_017025781.1 | 2244 | UTR 3 | XP_016881270.1 |