Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGGGTCATCCGACTGGGAGCTCC[A/C]GGAGTCCCGCCTGGTGGGGGGCGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TMEM200C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMEM200C - transmembrane protein 200C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080209.1 | 1608 | Missense Mutation | GGG,TGG | G,W 523 | NP_001073678.1 | |
XM_005258130.4 | 1608 | Missense Mutation | GGG,TGG | G,W 523 | XP_005258187.1 | |
XM_011525729.2 | 1608 | Missense Mutation | GGG,TGG | G,W 523 | XP_011524031.1 |