Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTTCTCCCCATGGTCAGCCACAC[A/G]TTCCACATGCGCACAGAGGAGTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TXNDC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TXNDC2 - thioredoxin domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098529.1 | 665 | Silent Mutation | ACA,ACG | T,T 72 | NP_001091999.1 | |
NM_032243.5 | 665 | Silent Mutation | ACA,ACG | T,T 5 | NP_115619.4 | |
XM_017026041.1 | 665 | Silent Mutation | ACA,ACG | T,T 5 | XP_016881530.1 |