Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAACTCTGGCATGAGGTTTACAGA[A/T]ATGTTTGTATACAACCTATCAACCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604017 | ||||||||||||||||||||
Literature Links: |
B4GALT6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
B4GALT6 - beta-1,4-galactosyltransferase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004775.3 | 1447 | Silent Mutation | ATA,ATT | I,I 368 | NP_004766.2 | |
XM_005258387.4 | 1447 | Silent Mutation | ATA,ATT | I,I 356 | XP_005258444.1 | |
XM_006722578.3 | 1447 | Silent Mutation | ATA,ATT | I,I 330 | XP_006722641.1 | |
XM_006722579.3 | 1447 | Silent Mutation | ATA,ATT | I,I 324 | XP_006722642.1 | |
XM_017026088.1 | 1447 | Silent Mutation | ATA,ATT | I,I 329 | XP_016881577.1 | |
XM_017026089.1 | 1447 | Silent Mutation | ATA,ATT | I,I 324 | XP_016881578.1 | |
XM_017026090.1 | 1447 | Silent Mutation | ATA,ATT | I,I 260 | XP_016881579.1 | |
XM_017026091.1 | 1447 | Silent Mutation | ATA,ATT | I,I 255 | XP_016881580.1 | |
XM_017026092.1 | 1447 | Silent Mutation | ATA,ATT | I,I 188 | XP_016881581.1 |