Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAATAAAAATGCAGCTGATTCTCG[A/G]TATCCTGCAACAAGCTGTCTCACGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603577 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NOL4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NOL4 - nucleolar protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198546.1 | 1886 | Silent Mutation | TAC,TAT | Y,Y 552 | NP_001185475.1 | |
NM_001198547.1 | 1886 | Silent Mutation | TAC,TAT | Y,Y 542 | NP_001185476.1 | |
NM_001198548.1 | 1886 | Silent Mutation | TAC,TAT | Y,Y 514 | NP_001185477.1 | |
NM_001198549.1 | 1886 | Silent Mutation | TAC,TAT | Y,Y 331 | NP_001185478.1 | |
NM_001282527.1 | 1886 | Silent Mutation | TAC,TAT | Y,Y 397 | NP_001269456.1 | |
NM_003787.4 | 1886 | Silent Mutation | TAC,TAT | Y,Y 616 | NP_003778.2 | |
XM_006722563.2 | 1886 | Silent Mutation | TAC,TAT | Y,Y 616 | XP_006722626.1 | |
XM_006722564.2 | 1886 | Silent Mutation | TAC,TAT | Y,Y 478 | XP_006722627.1 | |
XM_006722565.1 | 1886 | Silent Mutation | TAC,TAT | Y,Y 461 | XP_006722628.1 | |
XM_011526237.1 | 1886 | Silent Mutation | TAC,TAT | Y,Y 616 | XP_011524539.1 | |
XM_011526238.1 | 1886 | Silent Mutation | TAC,TAT | Y,Y 502 | XP_011524540.1 | |
XM_011526239.2 | 1886 | Silent Mutation | TAC,TAT | Y,Y 461 | XP_011524541.1 | |
XM_017026052.1 | 1886 | Silent Mutation | TAC,TAT | Y,Y 496 | XP_016881541.1 | |
XM_017026053.1 | 1886 | Silent Mutation | TAC,TAT | Y,Y 461 | XP_016881542.1 | |
XM_017026054.1 | 1886 | Silent Mutation | TAC,TAT | Y,Y 461 | XP_016881543.1 | |
XM_017026055.1 | 1886 | Silent Mutation | TAC,TAT | Y,Y 397 | XP_016881544.1 | |
XM_017026056.1 | 1886 | Silent Mutation | TAC,TAT | Y,Y 397 | XP_016881545.1 | |
XM_017026057.1 | 1886 | Silent Mutation | TAC,TAT | Y,Y 365 | XP_016881546.1 | |
XM_017026058.1 | 1886 | Silent Mutation | TAC,TAT | Y,Y 461 | XP_016881547.1 | |
XM_017026059.1 | 1886 | Silent Mutation | TAC,TAT | Y,Y 331 | XP_016881548.1 | |
XM_017026060.1 | 1886 | Silent Mutation | TAC,TAT | Y,Y 301 | XP_016881549.1 |