Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCCAATGAGATTCTCTTTCTCAA[A/C]TTTTGTTAATGGAAGATGATTCCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608731 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC39A6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC39A6 - solute carrier family 39 member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099406.1 | Intron | NP_001092876.1 | ||||
NM_012319.3 | Intron | NP_036451.3 | ||||
XM_011525900.1 | Intron | XP_011524202.1 | ||||
XM_011525901.1 | Intron | XP_011524203.1 |