Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTTGGGAGAGTAGGTTCGGCATC[A/G]GTCAACTGCTCCCTAGTGGGATCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615068 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPG5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EPG5 - ectopic P-granules autophagy protein 5 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020964.2 | Intron | NP_066015.2 | ||||
XM_011526122.1 | Intron | XP_011524424.1 | ||||
XM_017025889.1 | Intron | XP_016881378.1 | ||||
XM_017025890.1 | Intron | XP_016881379.1 | ||||
XM_017025891.1 | Intron | XP_016881380.1 | ||||
XM_017025892.1 | Intron | XP_016881381.1 | ||||
XM_017025893.1 | Intron | XP_016881382.1 |
SIGLEC15 - sialic acid binding Ig like lectin 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_213602.2 | Intron | NP_998767.1 |