Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATACACAATAGGAACCTGACCAAAT[G/T]AAGAAAAGAACAACTTACAATACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164360 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP5A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATP5A1 - ATP synthase, H+ transporting, mitochondrial F1 complex, alpha subunit 1, cardiac muscle | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001935.2 | Intron | NP_001001935.1 | ||||
NM_001001937.1 | Intron | NP_001001937.1 | ||||
NM_001257334.1 | Intron | NP_001244263.1 | ||||
NM_001257335.1 | Intron | NP_001244264.1 | ||||
NM_004046.5 | Intron | NP_004037.1 | ||||
XM_017025789.1 | Intron | XP_016881278.1 |