Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTTAAAGGAACAAAGCATTCTGT[C/T]TCCTTGCTTAGTACTGAGAACCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD29 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKRD29 - ankyrin repeat domain 29 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308238.1 | 1234 | UTR 3 | NP_001295167.1 | |||
NM_173505.3 | 1234 | UTR 3 | NP_775776.2 | |||
XM_011525830.2 | 1234 | UTR 3 | XP_011524132.1 | |||
XM_011525831.2 | 1234 | UTR 3 | XP_011524133.1 | |||
XM_011525832.2 | 1234 | UTR 3 | XP_011524134.1 | |||
XM_017025561.1 | 1234 | UTR 3 | XP_016881050.1 |