Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGCATCTTCAAGTAACCGGTAAGT[A/G]GCTCTTTCCTTTCTTCTCGTCGCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120470 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCC PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DCC - DCC netrin 1 receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005215.3 | Intron | NP_005206.2 | ||||
XM_011525843.1 | Intron | XP_011524145.1 | ||||
XM_011525844.2 | Intron | XP_011524146.1 | ||||
XM_017025568.1 | Intron | XP_016881057.1 | ||||
XM_017025569.1 | Intron | XP_016881058.1 | ||||
XM_017025570.1 | Intron | XP_016881059.1 |