Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTCTTGTGGAAACGTCCTCTTCA[A/G]TGGTTACCTGCCGCCACCAGGTAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606214 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SHKBP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SHKBP1 - SH3KBP1 binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138392.3 | 429 | Missense Mutation | AAT,AGT | N,S 127 | NP_612401.2 | |
XM_006723474.1 | 429 | Missense Mutation | AAT,AGT | N,S 127 | XP_006723537.1 | |
XM_006723475.1 | 429 | Missense Mutation | AAT,AGT | N,S 127 | XP_006723538.1 | |
XM_011527492.1 | 429 | Missense Mutation | AAT,AGT | N,S 127 | XP_011525794.1 | |
XM_011527493.1 | 429 | UTR 5 | XP_011525795.1 | |||
XM_017027473.1 | 429 | Missense Mutation | AAT,AGT | N,S 127 | XP_016882962.1 | |
XM_017027474.1 | 429 | UTR 5 | XP_016882963.1 |
SPTBN4 - spectrin beta, non-erythrocytic 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |