Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCATCCTGGAGGGGCGGTCTATGC[C/T]GAGAGCCCCACCAGCAGGAGGACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LYPD5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LYPD5 - LY6/PLAUR domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031749.2 | 881 | Silent Mutation | TCA,TCG | S,S 250 | NP_001026919.2 | |
NM_001288763.1 | 881 | Silent Mutation | TCA,TCG | S,S 207 | NP_001275692.1 | |
NM_182573.2 | 881 | Silent Mutation | TCA,TCG | S,S 207 | NP_872379.2 | |
XM_011526794.2 | 881 | Silent Mutation | TCA,TCG | S,S 207 | XP_011525096.1 |