Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATGATCGATTCAGGAGAAAAGAA[A/G]CCTGGGCGGAGAGCAGAGGTTTGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF222 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF222 - zinc finger protein 222 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001129996.1 | 137 | Silent Mutation | AAA,AAG | K,K 8 | NP_001123468.1 | |
NM_013360.2 | 137 | UTR 5 | NP_037492.2 | |||
XM_005259208.3 | 137 | UTR 5 | XP_005259265.1 | |||
XM_005259209.3 | 137 | Silent Mutation | AAA,AAG | K,K 8 | XP_005259266.1 | |
XM_017027237.1 | 137 | Silent Mutation | AAA,AAG | K,K 8 | XP_016882726.1 | |
XM_017027238.1 | 137 | UTR 5 | XP_016882727.1 |