Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTCAATAACTATCGCACAAAGCT[A/T]AGGCCACCAATGCTTTTATTTATCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 123310 MIM: 606495 | ||||||||||||||||||||
Literature Links: |
CKM PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CKM - creatine kinase, M-type | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001824.4 | 1657 | UTR 3 | NP_001815.2 | |||
XM_017026240.1 | 1657 | UTR 3 | XP_016881729.1 |
MARK4 - microtubule affinity regulating kinase 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |