Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGAGTGTACGCTGCACATCCCGG[C/T]GTCCTTTGAGCACATAGTTGGACCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604963 | ||||||||||||||||||||
Literature Links: |
CCDC61 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC61 - coiled-coil domain containing 61 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267723.1 | 786 | Intron | NP_001254652.1 | |||
XM_005259191.3 | 786 | Intron | XP_005259248.1 | |||
XM_005259192.4 | 786 | Intron | XP_005259249.1 | |||
XM_011527257.2 | 786 | Intron | XP_011525559.1 |
MIR769 - microRNA 769 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PGLYRP1 - peptidoglycan recognition protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005091.2 | 786 | Missense Mutation | CAC,CGC | H,R 170 | NP_005082.1 |