Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTGGACAAGGCCTCTATCATGCG[C/G]CTCACCATCAGCTACCTGCGCATGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609976 | ||||||||||||||||||||
Literature Links: |
HIF3A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HIF3A - hypoxia inducible factor 3 alpha subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022462.4 | 333 | Missense Mutation | GCC,GGC | A,G 38 | NP_071907.4 | |
NM_152794.3 | 333 | Silent Mutation | CGC,CGG | R,R 56 | NP_690007.1 | |
NM_152795.3 | 333 | Silent Mutation | CGC,CGG | R,R 58 | NP_690008.2 | |
NM_152796.2 | 333 | Missense Mutation | GCC,GGC | A,G 38 | NP_690009.1 | |
XM_005259152.4 | 333 | Silent Mutation | CGC,CGG | R,R 102 | XP_005259209.1 | |
XM_005259153.3 | 333 | Missense Mutation | GCC,GGC | A,G 38 | XP_005259210.2 | |
XM_005259155.4 | 333 | Silent Mutation | CGC,CGG | R,R 102 | XP_005259212.1 | |
XM_005259156.4 | 333 | Silent Mutation | CGC,CGG | R,R 102 | XP_005259213.1 | |
XM_017027132.1 | 333 | Silent Mutation | CGC,CGG | R,R 102 | XP_016882621.1 | |
XM_017027133.1 | 333 | Silent Mutation | CGC,CGG | R,R 102 | XP_016882622.1 | |
XM_017027134.1 | 333 | Silent Mutation | CGC,CGG | R,R 58 | XP_016882623.1 | |
XM_017027135.1 | 333 | Silent Mutation | CGC,CGG | R,R 56 | XP_016882624.1 | |
XM_017027136.1 | 333 | Silent Mutation | CGC,CGG | R,R 102 | XP_016882625.1 | |
XM_017027137.1 | 333 | Silent Mutation | CGC,CGG | R,R 56 | XP_016882626.1 | |
XM_017027138.1 | 333 | Silent Mutation | CGC,CGG | R,R 58 | XP_016882627.1 | |
XM_017027139.1 | 333 | Missense Mutation | GCC,GGC | A,G 38 | XP_016882628.1 | |
XM_017027140.1 | 333 | Missense Mutation | GCC,GGC | A,G 38 | XP_016882629.1 | |
XM_017027141.1 | 333 | Missense Mutation | GCC,GGC | A,G 38 | XP_016882630.1 | |
XM_017027142.1 | 333 | Missense Mutation | GCC,GGC | A,G 38 | XP_016882631.1 |