Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTCACACCCTGTCCCAGTCTCAC[A/G]CTTTCTGTGTCTCACGGTGGCATGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
GNG8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GNG8 - G protein subunit gamma 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033258.1 | 2091 | Intron | NP_150283.1 | |||
XM_011527524.2 | 2091 | Intron | XP_011525826.1 | |||
XM_017027503.1 | 2091 | Intron | XP_016882992.1 | |||
XM_017027504.1 | 2091 | UTR 5 | XP_016882993.1 | |||
XM_017027505.1 | 2091 | Intron | XP_016882994.1 |