Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCCTCGTTGAAAGGGAAGGTGCC[A/G]CTGAGGCTGACGTACATGATCACGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607074 | ||||||||||||||||||||
Literature Links: |
DACT3-AS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DACT3-AS1 - DACT3 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRKD2 - protein kinase D2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079880.1 | 2716 | Silent Mutation | AGC,AGT | S,S 746 | NP_001073349.1 | |
NM_001079881.1 | 2716 | Silent Mutation | AGC,AGT | S,S 746 | NP_001073350.1 | |
NM_001079882.1 | 2716 | Silent Mutation | AGC,AGT | S,S 589 | NP_001073351.1 | |
NM_016457.4 | 2716 | Silent Mutation | AGC,AGT | S,S 746 | NP_057541.2 | |
XM_005258716.2 | 2716 | Silent Mutation | AGC,AGT | S,S 589 | XP_005258773.2 |