Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATTGCAACCTCTGGCATAAATGG[A/G]TTAATGTTATTCCTTTTCTCATGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 180740 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SNRNP70 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SNRNP70 - small nuclear ribonucleoprotein U1 subunit 70 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301069.1 | Intron | NP_001287998.1 | ||||
NM_003089.5 | Intron | NP_003080.2 | ||||
XM_005259178.1 | Intron | XP_005259235.1 | ||||
XM_011527240.2 | Intron | XP_011525542.2 | ||||
XM_011527241.2 | Intron | XP_011525543.1 |