Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCAGGGGGCCAGGCTTATTCCCC[A/C]GGTCAGCCTCAAGGGCTTCTGGGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616633 MIM: 604429 | ||||||||||||||||||||
Literature Links: |
PRR12 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PRR12 - proline rich 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017027031.1 | 1813 | Silent Mutation | CCA,CCC | P,P 445 | XP_016882520.1 |
PRRG2 - proline rich and Gla domain 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |