Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGAAGTGCGTGGAGGTGACCAGG[G/T]TATAGAAGTTTCGGAGGAGGCGCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601026 MIM: 610622 MIM: 610197 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AP2A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AP2A1 - adaptor related protein complex 2 alpha 1 subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FUZ - fuzzy planar cell polarity protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171937.1 | 1147 | Missense Mutation | AAC,ACC | N,T 299 | NP_001165408.1 | |
NM_025129.4 | 1147 | Missense Mutation | AAC,ACC | N,T 335 | NP_079405.2 | |
XM_006723399.3 | 1147 | Missense Mutation | AAC,ACC | N,T 335 | XP_006723462.1 | |
XM_011527339.1 | 1147 | Missense Mutation | AAC,ACC | N,T 336 | XP_011525641.1 | |
XM_011527340.1 | 1147 | Missense Mutation | AAC,ACC | N,T 286 | XP_011525642.1 | |
XM_011527341.2 | 1147 | Missense Mutation | AAC,ACC | N,T 286 | XP_011525643.1 | |
XM_011527342.1 | 1147 | Missense Mutation | AAC,ACC | N,T 279 | XP_011525644.1 | |
XM_011527343.1 | 1147 | Missense Mutation | AAC,ACC | N,T 336 | XP_011525645.1 | |
XM_011527345.1 | 1147 | Missense Mutation | AAC,ACC | N,T 236 | XP_011525647.1 | |
XM_011527346.1 | 1147 | Missense Mutation | AAC,ACC | N,T 236 | XP_011525648.1 | |
XM_011527347.1 | 1147 | Missense Mutation | AAC,ACC | N,T 236 | XP_011525649.1 | |
XM_017027319.1 | 1147 | Missense Mutation | AAC,ACC | N,T 285 | XP_016882808.1 | |
XM_017027320.1 | 1147 | Missense Mutation | AAC,ACC | N,T 336 | XP_016882809.1 | |
XM_017027321.1 | 1147 | Missense Mutation | AAC,ACC | N,T 235 | XP_016882810.1 | |
XM_017027322.1 | 1147 | Missense Mutation | AAC,ACC | N,T 285 | XP_016882811.1 | |
XM_017027323.1 | 1147 | UTR 3 | XP_016882812.1 |
MED25 - mediator complex subunit 25 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |