Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCATGGTGTCATTTATCCACTTG[A/G]TGAACTTGCACACTTGAGTGTAGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602652 MIM: 604438 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLK6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KLK6 - kallikrein related peptidase 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KLK7 - kallikrein related peptidase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001207053.1 | 997 | Missense Mutation | NP_001193982.1 | |||
NM_001243126.1 | 997 | Missense Mutation | NP_001230055.1 | |||
NM_005046.3 | 997 | Missense Mutation | NP_005037.1 | |||
NM_139277.2 | 997 | Missense Mutation | NP_644806.1 |