Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGCGGGCTCAGTTCTCCATGATTT[C/T]TTGGATCCAGTCAAGGTAGTGGCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602673 MIM: 605644 MIM: 605504 | ||||||||||||||||||||
Literature Links: |
KLK10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KLK10 - kallikrein related peptidase 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KLK8 - kallikrein related peptidase 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KLK9 - kallikrein related peptidase 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012315.1 | 823 | Missense Mutation | AAA,GAA | K,E 246 | NP_036447.1 |