Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATGGGTCTGGAGAGAGAACATGC[A/G]TGCTGCAGGGTCCCCTAAATCTCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604434 MIM: 605539 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLK11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KLK11 - kallikrein related peptidase 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KLK12 - kallikrein related peptidase 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019598.2 | Intron | NP_062544.1 | ||||
NM_145894.1 | Intron | NP_665901.1 | ||||
NM_145895.1 | Intron | NP_665902.1 | ||||
XM_005258950.3 | Intron | XP_005259007.1 | ||||
XM_005258951.3 | Intron | XP_005259008.1 |