Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTTCAGTTGGTGTCAGAAATGAG[G/T]GGATCTTGTGGACCATTCCCTAACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SIGLECL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SIGLECL1 - SIGLEC family like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308437.1 | Intron | NP_001295366.1 | ||||
NM_173635.2 | Intron | NP_775906.1 | ||||
XM_005258799.4 | Intron | XP_005258856.1 | ||||
XM_011526821.2 | Intron | XP_011525123.1 | ||||
XM_017026647.1 | Intron | XP_016882136.1 |