Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCAAGAAAATGAAACAAATGGCCA[C/T]GAAGCACTCATGACAAAAACCAAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF701 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF701 - zinc finger protein 701 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172655.1 | 456 | Silent Mutation | CAC,CAT | H,H 175 | NP_001166126.1 | |
NM_018260.2 | 456 | Silent Mutation | CAC,CAT | H,H 109 | NP_060730.2 | |
XM_011527092.1 | 456 | Intron | XP_011525394.1 |