Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGTGAAGGAAGAGGGATGTATTG[C/T]GACCAAAGATCTTGCCACACTCATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF28 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF28 - zinc finger protein 28 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006969.3 | 1032 | Intron | NP_008900.3 | |||
XM_006723354.3 | 1032 | Intron | XP_006723417.1 | |||
XM_011527262.2 | 1032 | Missense Mutation | CAC,CGC | H,R 282 | XP_011525564.1 |