Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGCAAAGAGGGCCTGGGCCTGGC[C/T]CAGAAGGTAGATCCTGCCCAGAAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604910 | ||||||||||||||||||||
Literature Links: |
CNOT3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CNOT3 - CCR4-NOT transcription complex subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014516.3 | 691 | Silent Mutation | GCC,GCT | A,A 111 | NP_055331.1 | |
XM_005278279.1 | 691 | Silent Mutation | GCC,GCT | A,A 111 | XP_005278336.1 | |
XM_005278280.2 | 691 | Silent Mutation | GCC,GCT | A,A 111 | XP_005278337.1 | |
XM_005278281.1 | 691 | Silent Mutation | GCC,GCT | A,A 111 | XP_005278338.1 | |
XM_005278282.2 | 691 | Silent Mutation | GCC,GCT | A,A 111 | XP_005278339.1 | |
XM_011526992.1 | 691 | Silent Mutation | GCC,GCT | A,A 111 | XP_011525294.1 | |
XM_011526993.2 | 691 | Silent Mutation | GCC,GCT | A,A 111 | XP_011525295.1 |