Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCTGAGACTGGGCGGGCTCACTC[A/T]CCGCAGGACATAGGAACGGGCAGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606048 | ||||||||||||||||||||
Literature Links: |
MBOAT7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MBOAT7 - membrane bound O-acyltransferase domain containing 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146056.2 | Intron | NP_001139528.1 | ||||
NM_001146083.2 | Intron | NP_001139555.1 | ||||
NM_024298.4 | Intron | NP_077274.3 | ||||
XM_011527299.2 | Intron | XP_011525601.1 | ||||
XM_011527300.2 | Intron | XP_011525602.1 | ||||
XM_017027296.1 | Intron | XP_016882785.1 |
TMC4 - transmembrane channel like 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |