Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTGTCAGCAGTAAAACTTATATA[T/C]TTTTTAAAATAACTTCAATGTAGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604936 | ||||||||||||||||||||
Literature Links: |
KIR2DL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KIR2DL1 - killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014218.2 | 1515 | UTR 3 | NP_055033.2 | |||
XM_011526939.2 | 1515 | Intron | XP_011525241.1 | |||
XM_017026782.1 | 1515 | UTR 3 | XP_016882271.1 | |||
XM_017026783.1 | 1515 | Intron | XP_016882272.1 |
LOC102724300 - uncharacterized LOC102724300 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |