Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGACCAAGTTTCAGACAGAAAGA[G/T]CAAACTGGGTTTTCTTTCCTTTACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609664 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NLRP11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NLRP11 - NLR family pyrin domain containing 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297743.1 | 2790 | Missense Mutation | ATC,CTC | I,L 878 | NP_001284672.1 | |
NM_145007.3 | 2790 | Missense Mutation | ATC,CTC | I,L 977 | NP_659444.2 |