Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCTCATGGCAGCGGCGGTGCTGA[C/T]GGACCGGGTTCAGGTGAGTGGGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613840 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF304 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF304 - zinc finger protein 304 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290318.1 | 411 | Missense Mutation | ACG,ATG | T,M 7 | NP_001277247.1 | |
NM_001290319.1 | 411 | UTR 5 | NP_001277248.1 | |||
NM_020657.3 | 411 | Missense Mutation | ACG,ATG | T,M 7 | NP_065708.2 | |
XM_011527145.2 | 411 | UTR 5 | XP_011525447.1 | |||
XM_017027024.1 | 411 | UTR 5 | XP_016882513.1 |