Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGGCTCCGATGGCGGCGGCCGC[C/G]CTGAGGGACCCCGCTCAGGTGAGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF419 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF419 - zinc finger protein 419 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098491.1 | 255 | Silent Mutation | GCC,GCG | A,A 5 | NP_001091961.1 | |
NM_001098492.1 | 255 | Silent Mutation | GCC,GCG | A,A 5 | NP_001091962.1 | |
NM_001098493.1 | 255 | Silent Mutation | GCC,GCG | A,A 5 | NP_001091963.1 | |
NM_001098494.1 | 255 | Silent Mutation | GCC,GCG | A,A 5 | NP_001091964.1 | |
NM_001098495.1 | 255 | Silent Mutation | GCC,GCG | A,A 5 | NP_001091965.1 | |
NM_001098496.1 | 255 | Silent Mutation | GCC,GCG | A,A 5 | NP_001091966.1 | |
NM_001291743.1 | 255 | Silent Mutation | GCC,GCG | A,A 5 | NP_001278672.1 | |
NM_001291744.1 | 255 | UTR 5 | NP_001278673.1 | |||
NM_001291745.1 | 255 | Silent Mutation | GCC,GCG | A,A 5 | NP_001278674.1 | |
NM_024691.3 | 255 | Silent Mutation | GCC,GCG | A,A 5 | NP_078967.3 |
ZNF772 - zinc finger protein 772 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |