Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATTCAACTTGAGCGTGGCGAGGA[G/T]CCCTGGGTTCCCAGTGGAAAGGACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF324B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF324B - zinc finger protein 324B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207395.2 | 305 | Missense Mutation | GAG,GAT | E,D 58 | NP_997278.2 | |
XM_005258915.4 | 305 | Missense Mutation | GAG,GAT | E,D 58 | XP_005258972.1 | |
XM_005258918.4 | 305 | Missense Mutation | GAG,GAT | E,D 58 | XP_005258975.1 |