Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTTCAGTCTCTACAAAAATCTCA[C/T]TGTGGCTTCGAGGGCTTGTCTGTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603159 MIM: 607959 | ||||||||||||||||||||
Literature Links: |
LRP3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LRP3 - LDL receptor related protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002333.3 | 1351 | Intron | NP_002324.2 | |||
XM_005258945.1 | 1351 | Intron | XP_005259002.1 |
SLC7A10 - solute carrier family 7 member 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019849.2 | 1351 | Silent Mutation | CAA,CAG | Q,Q 523 | NP_062823.1 | |
XM_006723284.2 | 1351 | Silent Mutation | CAA,CAG | Q,Q 444 | XP_006723347.1 | |
XM_011527119.1 | 1351 | Silent Mutation | CAA,CAG | Q,Q 660 | XP_011525421.1 | |
XM_011527120.1 | 1351 | Silent Mutation | CAA,CAG | Q,Q 633 | XP_011525422.1 |