Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGCTGGAGGAGGATGAGCGCTTC[C/G]AGGGCCGCGTGGTGTGGAATGGCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 142440 MIM: 600235 | ||||||||||||||||||||
Literature Links: |
GRAMD1A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GRAMD1A - GRAM domain containing 1A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HPN - hepsin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCN1B - sodium voltage-gated channel beta subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037.4 | 632 | Missense Mutation | CAG,GAG | Q,E 87 | NP_001028.1 | |
NM_001321605.1 | 632 | Missense Mutation | CAG,GAG | Q,E 54 | NP_001308534.1 | |
NM_199037.4 | 632 | Missense Mutation | CAG,GAG | Q,E 87 | NP_950238.1 |