Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATGAGCATTCTGCAGCTCCTTCCC[A/G]GCCTGGCCAGCAGCATGGTGGGCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609169 MIM: 609969 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GAPDHS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GAPDHS - glyceraldehyde-3-phosphate dehydrogenase, spermatogenic | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SBSN - suprabasin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166034.1 | 1605 | Silent Mutation | GCC,GCT | A,A 523 | NP_001159506.1 | |
NM_001166035.1 | 1605 | Intron | NP_001159507.1 | |||
NM_198538.3 | 1605 | Silent Mutation | GCC,GCT | A,A 180 | NP_940940.1 | |
XM_011526929.2 | 1605 | Silent Mutation | GCC,GCT | A,A 505 | XP_011525231.1 | |
XM_011526930.2 | 1605 | Silent Mutation | GCC,GCT | A,A 505 | XP_011525232.1 | |
XM_011526931.2 | 1605 | Silent Mutation | GCC,GCT | A,A 487 | XP_011525233.1 |