Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGGGAGCCATTCACATACCCAGT[A/G]CTAGAGAGACGTGGTCACACATGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616377 | ||||||||||||||||||||
Literature Links: |
LINC01529 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LINC01529 - long intergenic non-protein coding RNA 1529 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRODH2 - proline dehydrogenase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021232.1 | 1296 | Missense Mutation | GCA,GTA | A,V 474 | NP_067055.1 | |
XM_017027084.1 | 1296 | Missense Mutation | GCA,GTA | A,V 398 | XP_016882573.1 | |
XM_017027085.1 | 1296 | Intron | XP_016882574.1 |