Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACGGAGGCAGGAATCGCCAACTGC[A/G]CCAGGCCTGAGGACACAGCGCGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607762 MIM: 602716 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIRREL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIRREL2 - kin of IRRE like 2 (Drosophila) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032123.6 | 221 | Intron | NP_115499.5 | |||
NM_199179.3 | 221 | Intron | NP_954648.3 | |||
NM_199180.3 | 221 | Intron | NP_954649.3 | |||
XM_011527362.1 | 221 | UTR 5 | XP_011525664.1 | |||
XM_011527363.1 | 221 | UTR 5 | XP_011525665.1 | |||
XM_011527365.1 | 221 | UTR 5 | XP_011525667.1 | |||
XM_011527366.1 | 221 | Intron | XP_011525668.1 | |||
XM_017027348.1 | 221 | Intron | XP_016882837.1 |
NPHS1 - NPHS1, nephrin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004646.3 | 221 | Missense Mutation | GCG,GTG | A,V 22 | NP_004637.1 |