Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGACATGCCAAATAAATTGTATG[A/G]CATAGTTCAAGGCTTTTCTAGATTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF790 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF790 - zinc finger protein 790 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242800.1 | 3193 | Intron | NP_001229729.1 | |||
NM_001242801.1 | 3193 | Intron | NP_001229730.1 | |||
NM_001242802.1 | 3193 | Intron | NP_001229731.1 | |||
NM_206894.3 | 3193 | Intron | NP_996777.2 | |||
XM_005258903.4 | 3193 | UTR 3 | XP_005258960.1 | |||
XM_011526950.2 | 3193 | UTR 3 | XP_011525252.1 |
ZNF790-AS1 - ZNF790 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |