Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTGGTTATCACAGGGAGCCAGTG[A/G]GAAGGCACCAGGTAAATTAGATTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF568 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF568 - zinc finger protein 568 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204835.1 | Intron | NP_001191764.1 | ||||
NM_001204836.1 | Intron | NP_001191765.1 | ||||
NM_001204837.1 | Intron | NP_001191766.1 | ||||
NM_001204838.1 | Intron | NP_001191767.1 | ||||
NM_001204839.1 | Intron | NP_001191768.1 | ||||
NM_198539.3 | Intron | NP_940941.2 | ||||
XM_017026772.1 | Intron | XP_016882261.1 | ||||
XM_017026773.1 | Intron | XP_016882262.1 | ||||
XM_017026774.1 | Intron | XP_016882263.1 | ||||
XM_017026775.1 | Intron | XP_016882264.1 | ||||
XM_017026776.1 | Intron | XP_016882265.1 | ||||
XM_017026777.1 | Intron | XP_016882266.1 | ||||
XM_017026778.1 | Intron | XP_016882267.1 | ||||
XM_017026779.1 | Intron | XP_016882268.1 | ||||
XM_017026780.1 | Intron | XP_016882269.1 |
ZNF829 - zinc finger protein 829 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |