Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAGGACTTTGATTTCTAAATGAA[C/T]CAGCACAGTCCAGCCCACCCAGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607430 MIM: 601240 MIM: 601825 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DAZAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DAZAP1 - DAZ associated protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GAMT - guanidinoacetate N-methyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000156.5 | 797 | Intron | NP_000147.1 | |||
NM_138924.2 | 797 | Missense Mutation | ATT,GTT | I,V 235 | NP_620279.1 |
NDUFS7 - NADH:ubiquinone oxidoreductase core subunit S7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |