Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTGAAGAGACGCGAGGGACGCCC[A/G]CGGCGCACCGCCGGCAGACGACGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608531 MIM: 602214 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BTBD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BTBD2 - BTB domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017797.3 | 1284 | UTR 3 | NP_060267.2 | |||
XM_005259593.1 | 1284 | UTR 3 | XP_005259650.1 | |||
XM_011528127.1 | 1284 | UTR 3 | XP_011526429.1 |
CSNK1G2 - casein kinase 1 gamma 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |