Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGGCCTTGGGGGGCTGGTCCGCGG[A/G]CAGGCAGGGCCCGCGGGGCTGGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608743 MIM: 601579 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C19orf35 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C19orf35 - chromosome 19 open reading frame 35 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
JSRP1 - junctional sarcoplasmic reticulum protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OAZ1 - ornithine decarboxylase antizyme 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301020.1 | Intron | NP_001287949.1 | ||||
NM_004152.3 | Intron | NP_004143.1 |